Anti-CaMKII beta Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88212
103274-660EA 734.58 USD
103274-660
Anti-CaMKII beta Rabbit Polyclonal Antibody
Antibodies
The CaMKII beta Antibody from Novus Biologicals is a rabbit polyclonal antibody to CaMKII beta. This antibody reacts with human, mouse. The CaMKII beta Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Simple Western

Type: Primary
Antigen: CaMKII beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR