Anti-CaMKII beta Mouse Monoclonal Antibody [clone: 2D7-D8]

H00000816-M02A
103323-088EA 583.08 USD
103323-088
Anti-CaMKII beta Mouse Monoclonal Antibody [clone: 2D7-D8]
The CaMKII beta Antibody (2D7-D8) from Novus Biologicals is a mouse monoclonal antibody to CaMKII beta. This antibody reacts with human. The CaMKII beta Antibody (2D7-D8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CaMKII beta
Clonality: Monoclonal
Clone: 2D7-D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR