Anti-CAM3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000808-B01P
103323-054EA 798.46 USD
103323-054
Anti-CAM3 Mouse Polyclonal Antibody
Antibodies
The Calmodulin 3 Antibody from Novus Biologicals is a mouse polyclonal antibody to Calmodulin 3. This antibody reacts with human. The Calmodulin 3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CAM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR