Anti-CALY Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91225
103280-706EA 734.58 USD
103280-706
Anti-CALY Rabbit Polyclonal Antibody
Antibodies
The CALY Antibody from Novus Biologicals is a rabbit polyclonal antibody to CALY. This antibody reacts with human. The CALY Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CALY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR