Anti-CALNB Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15664
102706-428EA 635.51 USD
102706-428
Anti-CALNB Rabbit Polyclonal Antibody
Antibodies
The Calcineurin B Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calcineurin B. This antibody reacts with human, mouse, rat. The Calcineurin B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CALNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR