Anti-CALN1 Mouse Polyclonal Antibody

H00083698-A01
103344-878EA 436.92 USD
103344-878
Anti-CALN1 Mouse Polyclonal Antibody
The Calneuron 1 Antibody from Novus Biologicals is a mouse polyclonal antibody to Calneuron 1. This antibody reacts with human. The Calneuron 1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CALN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR