Anti-CALML5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15668
102706-436EA 635.51 USD
102706-436
Anti-CALML5 Rabbit Polyclonal Antibody
Antibodies
The CALML5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CALML5. This antibody reacts with human. The CALML5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CALML5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR