Anti-CALML5 Mouse Monoclonal Antibody [clone: 2F10]

Supplier: Novus Biologicals
H00051806-M16
103344-534EA 798.46 USD
103344-534
Anti-CALML5 Mouse Monoclonal Antibody [clone: 2F10]
Antibodies
The CALML5 Antibody (2F10) from Novus Biologicals is a mouse monoclonal antibody to CALML5. This antibody reacts with human. The CALML5 Antibody (2F10) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CALML5
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR