Anti-CALML3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000810-B02P
103323-058EA 399 USD
103323-058
Anti-CALML3 Mouse Polyclonal Antibody
Antibodies
The CALML3 Antibody from Novus Biologicals is a mouse polyclonal antibody to CALML3. This antibody reacts with human. The CALML3 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CALML3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR