Anti-CALD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
22380002-0.1MG
103317-748EA 399 USD
103317-748
Anti-CALD1 Rabbit Polyclonal Antibody
The Caldesmon / CALD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Caldesmon / CALD1. This antibody reacts with human. The Caldesmon / CALD1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CALD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR