Anti-CACYBP Mouse Monoclonal Antibody [clone: 2000]

Supplier: Novus Biologicals
H00027101-M01
103342-574EA 798.46 USD
103342-574
Anti-CACYBP Mouse Monoclonal Antibody [clone: 2000]
Antibodies
The CACYBP Antibody (2E3) from Novus Biologicals is a mouse monoclonal antibody to CACYBP. This antibody reacts with human. The CACYBP Antibody (2E3) has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.

Proximity Ligation Assay

Type: Primary
Antigen: CACYBP
Clonality: Monoclonal
Clone: 2000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR