Anti-CACYBP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00027101-D01P
103342-572EA 429 USD
103342-572
Anti-CACYBP Rabbit Polyclonal Antibody
Antibodies
The CACYBP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACYBP. This antibody reacts with human. The CACYBP Antibody has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.

Proximity Ligation Assay

Type: Primary
Antigen: CACYBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR