Anti-CACNB4 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1485
103368-262EA 329 USD
103368-262
Anti-CACNB4 Goat Polyclonal Antibody
Antibodies
The CACNB4 Antibody from Novus Biologicals is a goat polyclonal antibody to CACNB4. This antibody reacts with human. The CACNB4 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Peptide

Type: Primary
Antigen: CACNB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR