Anti-CACNB3 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00000784-B01P
103323-022EA 798.46 USD
103323-022
Anti-CACNB3 Mouse Polyclonal Antibody
Antibodies
The CACNB3 Antibody from Novus Biologicals is a mouse polyclonal antibody to CACNB3. This antibody reacts with human. The CACNB3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CACNB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR