Anti-CACNB2 Mouse Monoclonal Antibody [clone: 6C1]

Supplier: Novus Biologicals
H00000783-M01
103323-012EA 798.46 USD
103323-012
Anti-CACNB2 Mouse Monoclonal Antibody [clone: 6C1]
Antibodies
The CACNB2 Antibody (6C1) from Novus Biologicals is a mouse monoclonal antibody to CACNB2. This antibody reacts with human, mouse, rat. The CACNB2 Antibody (6C1) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CACNB2
Clonality: Monoclonal
Clone: 6C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR