Anti-CACNB1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00000782-D01P
103323-008EA 844.62 USD
103323-008
Anti-CACNB1 Rabbit Polyclonal Antibody
Antibodies
The CACNB1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACNB1. This antibody reacts with human. The CACNB1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CACNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR