Anti-CACNA2D4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-85920
103272-266EA 734.58 USD
103272-266
Anti-CACNA2D4 Rabbit Polyclonal Antibody
Antibodies
The CACNA2D4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACNA2D4. This antibody reacts with human. The CACNA2D4 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CACNA2D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR