Anti-CACNA2D2 Mouse Monoclonal Antibody [clone: 4000]

Supplier: Novus Biologicals
H00009254-M12
103336-544EA 798.46 USD
103336-544
Anti-CACNA2D2 Mouse Monoclonal Antibody [clone: 4000]
Antibodies
The CACNA2D2 Antibody (4E3) from Novus Biologicals is a mouse monoclonal antibody to CACNA2D2. This antibody reacts with human. The CACNA2D2 Antibody (4E3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CACNA2D2
Clonality: Monoclonal
Clone: 4000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR