Anti-CACNA1S Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-33799
103283-232EA 734.58 USD
103283-232
Anti-CACNA1S Rabbit Polyclonal Antibody
Antibodies
The CACNA1S Antibody from Novus Biologicals is a rabbit polyclonal antibody to CACNA1S. This antibody reacts with human. The CACNA1S Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CACNA1S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR