Anti-CAB39 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00051719-B01P
103344-414EA 798.46 USD
103344-414
Anti-CAB39 Mouse Polyclonal Antibody
Antibodies
The CAB39 Antibody from Novus Biologicals is a mouse polyclonal antibody to CAB39. This antibody reacts with human. The CAB39 Antibody has been validated for the following applications: Western Blot, ELISA, Proximity Ligation Assay.

Proximity Ligation Assay

Type: Primary
Antigen: CAB39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR