Anti-CA9 Rabbit Polyclonal Antibody (Alexa Fluor® 405)

Supplier: Novus Biologicals
NB100-417AF405
103358-002EA 688 USD
103358-002
Anti-CA9 Rabbit Polyclonal Antibody (Alexa Fluor® 405)
Antibodies
The Carbonic Anhydrase IX / CA9 Antibody [Alexa Fluor« 405] from Novus Biologicals is a rabbit polyclonal antibody to Carbonic Anhydrase IX / CA9. This antibody reacts with human, mouse, rat, canine, plant. The Carbonic Anhydrase IX / CA9 Antibody [Alexa Fluor« 405] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR