Anti-CA9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Novus Biologicals
NB100-417F
103358-010EA 688 USD
103358-010
Anti-CA9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The Carbonic Anhydrase IX / CA9 Antibody [FITC] from Novus Biologicals is a rabbit polyclonal antibody to Carbonic Anhydrase IX / CA9. This antibody reacts with human, mouse, rat, canine, plant. The Carbonic Anhydrase IX / CA9 Antibody [FITC] has been validated for the following applications: Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine, Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR