Anti-CA5B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86090
103272-520EA 734.58 USD
103272-520
Anti-CA5B Rabbit Polyclonal Antibody
Antibodies
The Carbonic Anhydrase VB / CA5B Antibody from Novus Biologicals is a rabbit polyclonal antibody to Carbonic Anhydrase VB / CA5B. This antibody reacts with human, rat. The Carbonic Anhydrase VB / CA5B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CA5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR