Anti-CA125/MUC16 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-52163
103408-320EA 299 USD
103408-320
Anti-CA125/MUC16 Goat Polyclonal Antibody
Antibodies
The CA125 / MUC16 Antibody from Novus Biologicals is a goat polyclonal antibody to CA125 / MUC16. This antibody reacts with human. The CA125 / MUC16 Antibody has been validated for the following applications: Peptide ELISA.

Peptide

Type: Primary
Antigen: CA125/MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-PQNLSIRGEYQIN
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR