Anti-CA125/MUC16 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
30400002-0.1MG
103317-156EA 349 USD
103317-156
Anti-CA125/MUC16 Rabbit Polyclonal Antibody
Antibodies
The CA125 / MUC16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CA125 / MUC16. This antibody reacts with human. The CA125 / MUC16 Antibody has been validated for the following applications: ELISA.

Type: Primary
Antigen: CA125/MUC16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR