Anti-CA1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00000759-D01P
103322-956EA 844.62 USD
103322-956
Anti-CA1 Rabbit Polyclonal Antibody
Antibodies
The Carbonic Anhydrase I / CA1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Carbonic Anhydrase I / CA1. This antibody reacts with human, mouse. The Carbonic Anhydrase I / CA1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR