Anti-C9orf98 Mouse Monoclonal Antibody [clone: 3B8]

Supplier: Novus Biologicals
H00158067-M01
103347-432EA 798.46 USD
103347-432
Anti-C9orf98 Mouse Monoclonal Antibody [clone: 3B8]
Antibodies
The C9orf98 Antibody (3B8) from Novus Biologicals is a mouse monoclonal antibody to C9orf98. This antibody reacts with human. The C9orf98 Antibody (3B8) has been validated for the following applications: Western Blot, ELISA, RNA Inhibition.

RNA Inhibition

Type: Primary
Antigen: C9orf98
Clonality: Monoclonal
Clone: 3B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR