Anti-C9orf5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90633
103280-470EA 734.58 USD
103280-470
Anti-C9orf5 Rabbit Polyclonal Antibody
Antibodies
The C9orf5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to C9orf5. This antibody reacts with human. The C9orf5 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: C9orf5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR