You Searched For: N,N,N',N'-Tetramethylethylenediamine (TEMED)


14  results were found

SearchResultCount:"14"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: N, N, N', N'-Tetramethyl ethylenediamine for synthesis, CAS number 110-18-9, Chemical formula (CH3)2NCH2CH2N(CH3)2, Synonyms TEMED, TMEDA, 1, 2-Bis(dimethylamino)ethane 5ml
Supplier: MP Biomedicals
Description: N,N,N',N'-Tetramethylethylenediamin is a tertiary amine base.

SDS

Supplier: Thermo Scientific Chemicals
Description: NNNN-TETRAMETHYLETHYLENEDIAMI 2.5L

Supplier: TCI America
Description: , 110-18-9, C6H16N2, 116.21
Supplier: Thermo Scientific Chemicals
Description: 99%
Supplier: MP Biomedicals
Description: N,N,N',N'-Tetramethylethylenediamin is a tertiary amine base.
Supplier: Thermo Scientific Chemicals
Description: NNNNTETRAMETHYLETHYLENEDIAMINE 99% 500G

Supplier: TCI America
Description: CAS Number: 110-18-9 MDL Number: MFCD00008335 Molecular Formula: C6H16N2 Molecular Weight: 116.21 Purity/Analysis Method: 98.0% (GC,T) Form: ...
Catalog Number: (TS43383-1000)
Supplier: Thermo Scientific Chemicals
Description: NNNN-TETRAMETHYLETHYLENEDIAMI 100ML


Supplier: G-Biosciences
Description: G-Biosciences offers TEMED (N,N,N’,N’-Tetramethylethylenediamine), a polymerization reagent used when making polyacrylamide gels used in gel electroph...

SDS

Supplier: BeanTown Chemical
Description: N, N, N', N'-Tetramethylethylenediamine, Purity: 99%, CAS Number: 110-18-9, Molecular Formula: C6H16N2, Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, Size: 500ML

SDS

Supplier: Thermo Scientific Chemicals
Description: Liquid
Supplier: APOLLO SCIENTIFIC
Description: NNNN-TETRAMETHYLETHANE-1 2-DIAMINE 250ML

New Product

Supplier: Strem Chemicals Inc
Description: CAS #: 110-18-9. Size: 500g.

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 14 of 14
no targeter for Bottom