You Searched For: Ammonium iron (III) sulphate dodecahydrate


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Crystals. Lot analysis on label.
Catalog Number: (97061-254)
Supplier: VWR
Description: FERRIC AMM SO4 DODECAHYDRATE ACS 500G

Supplier: MilliporeSigma
Description: AMMONIUM IRON(III) SULFATE DOD
Supplier: Thermo Scientific Chemicals
Description: 2.5KG
Catalog Number: (RC3058-32)
Supplier: Ricca Chemical
Description: FERRIC AMMONIUM SULFATE TS

Catalog Number: (BJ31111-500G)
Supplier: Honeywell Research Chemicals
Description: Ammonium iron(III) sulfate dodecahydrate, Purity: 99.0-102.0%, 99.0-102.0%, Grade: ACS, CAS Number: 7783-83-7, Molecular formula: NH4Fe(SO4)2.12H2O, Molar mass: 40.00 g/mol, Container Type: Poly bottle, Appearance: slightly violet, Size: 500G

SDS


Supplier: Thermo Scientific Chemicals
Description: AMMONIUM IRON(III) SULFATE DODECAH 500G

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM IRON(III) SULFATE DODECAH 500GR

Supplier: Thermo Scientific Chemicals
Description: In dyeing and printing textiles, as analytical lab reagent
Catalog Number: (101410-652)
Supplier: Electron Microscopy Sciences
Description: Electron Microscopy Sciences Ferric Ammonium Sulfate is available in concentrations of 4.0% aqueous and 5.0% aqueous.

Minority or Woman-Owned Business Enterprise


Supplier: BeanTown Chemical
Description: Ammonium iron(III) sulfate dodecahydrate, ACS grade, Purity: 95.8-102.0%, CAS No. 7783-83-7, Molecular Formula: H4FeNO8S2.12H2O, Synonyms: Ammonium ferric sulfate, Ferric ammonium sulfate, Size: 2.5KG

SDS

Catalog Number: (101410-654)
Supplier: Electron Microscopy Sciences
Description: Electron Microscopy Sciences Ferric Ammonium Sulfate is available in concentrations of 4.0% aqueous and 5.0% aqueous.

Minority or Woman-Owned Business Enterprise


Catalog Number: (76697-598)
Supplier: Ambeed
Description: AMMONIUM IRON SULFATE DODECAHYDRATE 100G

New Product


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom