di-Ammonium oxalate monohydrate
Catalog Number:
(97064-792)
Catalog Number:
(JT0746-R)
Supplier:
MilliporeSigma
Description:
EMSURE* di-Ammonium oxalate monohydrate for analysis 250gm^
Supplier:
MilliporeSigma
Description:
Di-Ammonium oxalate monohydrate, extra pure, CAS number: 6009-70-7, 50kg^
Supplier:
Alfa Aesar
Description:
Beilstein Registry No.: 3759830
Common Applications: In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals...
Supplier:
BeanTown Chemical
Description:
CAS: 6009-70-7; EC No: 214-202-3; MDL No: MFCD00149694
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Granular; Linear Formula: (NH4)C2O4·H2O; MW: ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the
![]()
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
* Please note if your account is within the State of California two of these pieces of identification will be required.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
|
|||||||||