You Searched For: Sodium hypochlorite


17  results were found

SearchResultCount:"17"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (BDH7038-4L)
Supplier: VWR International
Description: SODIUM HYPOCHLORITE 10-14 % W/W

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Sodium Hypochlorite Solution (5% Available Chlorine), BAKER ANALYZED® Reagent
Catalog Number: (JTP005-3)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: BAKER - A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by ...

Supplier: Ricca Chemical
Description: SODIUM HYPOCHLORITE SOLUTION CLRN 1L
Supplier: Ricca Chemical
Description: SODIUM HYPOCHLORITE SOL 6% CLRN 500ML
Supplier: Thermo Scientific Chemicals
Description: SODIUM HYPOCHLORITE 500ML

Supplier: Ricca Chemical
Description: approx. 2.5% NaOCl. APHA and EPA for Ammonia Nitrogen. Container: Plastic.
Supplier: Thermo Scientific Chemicals
Description: Typically 12 to 14% NaOCl
Supplier: Honeywell Research Chemicals
Description: Sodium hypochlorite solution available chlorine 4.00-4.99%, Grade: Reagent, CAS Number: 7681-52-9, Molecular formula: NaClO, Molar mass: 74.44 g/mol, Container Type: Poly bottle, Appearance: Colorless to Yellow, Application: Laboratory Chemical, Size: 500ML

Catalog Number: (BJ71696-10L)
Supplier: Honeywell Research Chemicals
Description: Sodium hypochlorite solution, General Purpose Reagent, Synthesis, Laboratory Chemical, Purity: approx 10% (RT), Grade: Purum, CAS no: 7681-52-9, MF: NaClO, Molar Mass: 74.44 g/mol, Appearance: Slightly Yellow To Deep Gre

Supplier: Honeywell Research Chemicals
Description: Sodium hypochlorite solution, Grade: reagent, Available chlorine 10-15%, CAS Number: 7681-52-9, Molecular formula: NaClO, Molar Weight: 74.44 g/mol, Container: Poly Bottle, Appearance: Faint Yellow To Dark Yellow, Form: Li
Supplier: Honeywell Research Chemicals
Description: Sodium hypochlorite solution, Cas number: 7681-52-9, Linear Formula: NaClO, Molar Mass: 74.44 g/mol, 6-14% active chlorine basis, Size: 6X500ML

SDS

Catalog Number: (700001-340)
Supplier: Spectrum Chemicals
Description: Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solut...

Catalog Number: (700001-334)
Supplier: Spectrum Chemicals
Description: Sodium Hypochlorite, 5 Percent Available Chlorine, Solution, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide soluti...

Supplier: Thermo Scientific Chemicals
Description: SODIUM HYPOCHLORITE 2.5L

Supplier: Hawkins Pharmaceutical Group
Description: Extremely low mercury.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 17
no targeter for Bottom