Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Sale Items

Classroom Furniture

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"27"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  TRIUMPH BOARD A.S.
Description:   Triumph Board Interactive Flat Panels are fully integrated, professional, multi-touch UHD displays with amazing performance and stunning visuals. They...
Supplier:  TRIUMPH BOARD A.S.
Description:   Over 4,500 Curriculum-Based Resources Including Over 70 Videos, Multitouch, Record Sound And Video, Text-To-Speach Capabilities, Widget Bank, Language Tools, Numeracy Tools, Content Packs, And Much More
Catalog Number: (470019-144)

Supplier:  TRIUMPH BOARD A.S.
Description:   Eliminates Shadow And Glare Due To Close Proximity Of Projector To Board. Offering 2D And 3D Projection. Native Resolution Wxga 1280 X 800. Brightness 3500 Ansi Lumens, Contrast 15000:1
Supplier:  TRIUMPH BOARD A.S.
Description:   Voting System (25 Clickers+1 Teacher Clicker and Receiver) with LCD Display, Carry Case, Installation Instructions, Triumph Board Vote Software Cd (Site Licence). Audience Response/Voting System with RF Technology Included Editor Software.
Catalog Number: (470019-184)

Supplier:  TRIUMPH BOARD A.S.
Description:   With Ir Camera And Laser Curtain Modules, Laser Curtain Ir Laser 830Nm, 20,000 Hours Lifetime, Image Size Support 75-100, Number Of T Ouch Points 10 Multi-Touch Points, Auto Calibration / Manual 4 Points Calibration
Catalog Number: (470019-472)

Supplier:  TRIUMPH BOARD A.S.
Description:   Triumph Board Stylus (White)^
Catalog Number: (470019-120)

Supplier:  TRIUMPH BOARD A.S.
Description:   Eliminates Shadow And Glare Due To Close Proximity Of Projector To Board. Offering 2D And 3D Projection. Native Resolution Xga 1024 X 768. Brightness 3000 Ansi Lumens, Contrast 15000:1, Noise Level 30Db (Eco Mode), ), Aspect Ratio 4:3 Native, Weight: \<7.5Kg
Catalog Number: (470214-354)

Supplier:  TRIUMPH BOARD A.S.
Description:   Voting System (25 Clickers+1 Teacher Clicker and Receiver), Carry Case, Installation Instructions, Triumph Board Vote Software Cd (Site Licence) Audience Response/Voting System with RF Technology Included Editor Software or Simply Integrate Voting
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 27
  1  2  3  4  Next