Chloroform : isoamyl alcohol (24:1) % ≥99% with red dye, Ready-Red™ for molecular biology
11RYRED001 11RYRED002
76180-748PK 424.89 USD
76180-748 76180-746
Chloroform : isoamyl alcohol (24:1) % ≥99% with red dye, Ready-Red™ for molecular biology
Chloroform : isoamyl alcohol
Ready-Red™ is a Chloroform:isoamyl alcohol 24:1 with red dye solution which facilitates separation of organic and aqueous phases.
  • Molecular Biology Grade
  • Purity >99%
  • Improves Recovery
Ready-Red™ is used either to remove traces of phenol from the aqueous phase or can be mixed directly with AquaPhenol™ (void volume in AquaPhenol™ bottle is adapted to this purpose).
Ready-Red™ is a coloured mixture of chloroform and isoamyl alcohol. The red colour (non toxic dye), facilitates the visualization and the separation of aqueous and organic phases during nucleic acids purification by means of phenolic extractions. The red dye of Ready-Red™ is not water-soluble and does not interfere with the quality of the nucleic acids purified. The nucleic acids recovered can be used in all standard experiments of molecular biology such as sequencing, PCR, cloning, transformations, etc.