Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "iodine-concentrated-aqueous-solution"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Honeywell Research Chemicals
Description:   Iodine concentrate, Grade: Analytical, Cas number: 7553-56-2, Molecular Formula: I2, Molar mass: 60.05 g/mol, Quality: For 1L standard solution, Concentration: 0.05 M I2, Synonym: Iodine solution, Volumetric Concentrate, Container: Ampoule, Size: 1l