    Sort by:
  • Search Within Results
2,5-Diphenylfuran
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (TCD3176-1G)
Supplier:  TCI America
Description:   CAS Number: 955-83-9 Molecular Formula: C16H12O Molecular Weight: 220.27 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C):...