Proheat Varitemp Heat Gun, Master Appliance
Supplier: Master Appliance
Proheat®
467-PH-1200
76006-434EA 474.51 USD
76006-434
Proheat Varitemp Heat Gun, Master Appliance
Heaters Hot Air Dryers
An Off/Cool/Hot switch allows users to cool down heating element after use, eliminating residual heat
  • Lightweight, compact, and easy-to-use with built-in stand
  • Ergonomic design for superb balance and grip
  • Dial-in variable temperature control and variable air flow
  • Single fan speed
  • 4000 ft./min. velocity
  • Made in USA
Special electronic closed loop temperature control system maintains constant temperature and protects unit from overheating. Equipped with a pistol grip handle and 6' cord.
Use to desolder, heat temperature-sensitive materials, shrink tubing, terminate specialty connectors, and more.
Certifications: ESD Safe; UL and cUL Listed.