You Searched For: Potassium hydroxide


141  results were found

SearchResultCount:"141"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (BDH7730-1)
Supplier: VWR International
Description: Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Supplier: VWR International
Description: POTASSIUM HYDROXIDE 45% W/W 1L
Catalog Number: (BDH7622-1)
Supplier: VWR International
Description: APHA for anaerobic sludge.

Supplier: VWR International
Description: Meets reagent specifications for testing USP/NF monographs.
Catalog Number: (BDH7907-1)
Supplier: VWR International
Description: Made with deionized water.

Catalog Number: (BDH7630-1)
Supplier: VWR International
Description: POTASSIUM HYDROXIDE 0.1N 1L

Catalog Number: (BDH7211-1)
Supplier: VWR International
Description: Made with deionized water. Suitable as a titrant.

Supplier: VWR International
Description: POTASSIUM HYDROXIDE 0.1N 4L
Catalog Number: (BDH7212-1)
Supplier: VWR International
Description: Made with deionized water. Suitable as a titrant.

Catalog Number: (BDH7457-1)
Supplier: VWR International
Description: Made with 200 proof ethyl alcohol. Prepared to ASTM D 94.

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Potassium Hydroxide, Pellets, N.F. - F.C.C.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Potassium Hydroxide, Pellets, AR® (ACS)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Standardized at 25 °C against chemicals whose certification is traceable to NIST.
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Potassium Hydroxide, Pellets, BAKER ANALYZED® Electronic Grade
Catalog Number: (JT3144-3)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Potassium Hydroxide, 45%, BAKER ANALYZED® Electronic Grade

Supplier: VWR
Description: POTASSIUM HYDROXIDE REAGENT GRADE 2.5KG
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 141
no targeter for Bottom