You Searched For: Dimethyl sulphoxide


60  results were found

SearchResultCount:"60"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR International
Description: BDH Dimethyl Sulfoxide Is Suitable For General Laboratory Use Including Cytology, Histology, Organic Synthesis, Cleaning, Prep Scale Synthesis. Packaged 4-Liter Plastic Bottles, With 4 Bottles Per Case. ACS Certified.
Catalog Number: (BDH67001.400)
Supplier: VWR International
Description: BDH* Dimethyl Sulfoxide, Chemical Formula: (CH3)2SO, Melting Point: 18.5, Boiling Point: 189, Flash Point: 95 deg C, Formula Weight: 78.14, CAS Number: 67-68-5, Grade: HPLC/Spectro, HiPerSolv CHROMANORM, Class: F, Size: 4L

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Dimethyl Sulfoxide, Multi-Compendial
Catalog Number: (MK296908)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Suitable for Liquid Chromatography and UV-Spectrophotometry.

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Dimethyl Sulfoxide, BAKER ANALYZED® A.C.S. Reagent
Supplier: VWR
Description: DIMETHYL SULFOXIDE ACS GRADE 4L
Catalog Number: (97063-136)
Supplier: VWR
Description: DIMETHYL SULFOXIDE ULT-PU GRADE 5X10ML

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Dimethyl Sulfoxide, AR® (ACS)
Catalog Number: (JT9194-33)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: For spectrophotometry. UV cutoff 270 nm. Lot analysis on label.

Supplier: MilliporeSigma
Description: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Supplier: MilliporeSigma
Description: These reagents are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency. To pro...
Supplier: MilliporeSigma
Description: Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents ran...
Supplier: MilliporeSigma
Description: Clear liquid. For HPLC, gas chromatography, and spectrophotometry. The infrared spectrum conforms to the standard. Filtered through a 0.2µm element. U...
Supplier: MilliporeSigma
Description: DIMETHYL SULFOXIDE FOR (2.5L)
Supplier: MilliporeSigma
Description: SupraSolv® solvents for headspace gas chromatography are developed particularly for the analysis of residual solvents in drug substances, excipients a...
Catalog Number: (EM1.09678.0100)
Supplier: MilliporeSigma
Description: DIMETHYL SULFOXIDE FOR (100ML)

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 60
no targeter for Bottom