    Sort by:
  • Search Within Results
Iodoacetamide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"11"
  List View Easy View
Sort by:
Catalog Number: (97064-926)
Supplier:  VWR
Description:   Iodoacetamide Proteomics Grade^
SDS
Supplier:  MilliporeSigma
Description:   2-Iodoacetamide for synthesis, CAS number 144-48-9, Chemical formula ICH2CONH2 25g^
SDS
Supplier:  Alfa Aesar
Description:   Alkylating agent used in peptide mapping because it covalently binds with the thiols in cysteine to prevent disulfide bond formation
SDS
Supplier:  TCI America
Description:   CAS Number: 144-48-9 MDL Number: MFCD00008028 Molecular Formula: C2H4INO Molecular Weight: 184.96 Purity/Analysis Method: 98.0% (N) Form: Cry...
SDS
Supplier:  TCI America
Description:   [for Biochemical Research] CAS Number: 144-48-9 MDL Number: MFCD00008028 Molecular Formula: C2H4INO Molecular Weight: 184.96 Purity/Analysis Metho...
SDS
Supplier:  MP Biomedicals
Description:   Iodoacetamide is a thiol reagent; alkylating reagent for cysteine and histidine residues in proteins. In the alkylation reaction, it reacts with histi...
SDS
Catalog Number: (82023-296)
Supplier:  G-Biosciences
Description:   OneQuant™ iodoacetamide are single aliquots of iodoacetamide that eliminate the need for weighing, preventing loss of reagent and saving time. Add 15...
SDS
Supplier:  G-Biosciences
Description:   G-Biosciences offers proteomic grade iodoacetamide in two bulk quantities (5 or 25 grams) or in OneQuant™ format. G-Biosciences' OneQuant™ Iodoacetami...
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next 