You Searched For: Guanidinium thiocyanate


15  results were found

SearchResultCount:"15"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR
Description: A powerful protein denaturant used to inactivate endogenous RNases during the isolation of RNA from tissues and bacterial cultures
Supplier: MilliporeSigma
Description: GUANIDINIUM THIOCYANATE (250G)
Supplier: TCI America
Description: CAS Number: 593-84-0 MDL Number: MFCD00013027 Molecular Formula: CH5N3 Molecular Weight: 118.16 Purity/Analysis Method: 99.0% (T) Form: Cryst...
Supplier: Thermo Scientific Chemicals
Description: GUANIDINE THIOCYANATE 1KG

Supplier: TCI America
Description: [for Biochemical Research] CAS Number: 593-84-0 MDL Number: MFCD00013027 Molecular Formula: CH5N3 Molecular Weight: 118.16 Purity/Analysis Method:...
Supplier: Thermo Scientific Chemicals
Description: 99% 500G
Catalog Number: (TS43394-1000)
Supplier: Thermo Scientific Chemicals
Description: GUANIDINE THIOCYANATE 100G


Catalog Number: (IC820991)
Supplier: MP Biomedicals
Description: Guanidine Thiocyanate is a chaotropic agent and strong protein denaturant which inactivates nucleases approximately 2.5 times faster than guanidine hy...

SDS


Supplier: MilliporeSigma
Description: Powerful protein denaturant widely used in purification of proteins and nucleic acids.
Supplier: MilliporeSigma
Description: For molecular biology. DNase-, RNase-, and protease-free.
Supplier: BeanTown Chemical
Description: Guanidine thiocyanate, Purity: 99%, CAS no. 593-84-0, Chemical Formula: C2H6N4S, Synonym: Guanidinium rhodanide, Size: 500G

SDS

Supplier: G-Biosciences
Description: Molecular molecular weight: 118.16

SDS

Supplier: Ambeed
Description: Guanidine Thiocyanate, Purity: 99%, CAS Number: 593-84-0, Appearance: Form: Crystal - Powder / Colour: White - Almost white, Storage: Keep in dark place, Inert atmosphere, Room temperature, Size: 100g

New Product

Supplier: Thermo Scientific Chemicals
Description: A protein denaturant
Catalog Number: (IB05100)
Supplier: IBI Scientific
Description: Guanidine Thiocyanate is used in the isolation of RNA.

SDS


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 15 of 15
no targeter for Bottom