You Searched For: Ammonium fluoride


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: AMMONIUM FLUORIDE FOR ANALYSIS
Supplier: Thermo Scientific Chemicals
Description: 97% 500G
Supplier: Thermo Scientific Chemicals
Description: For etching glass, in printing textile, for preservation of wood
Supplier: Spectrum Chemicals
Description: Ammonium Fluoride, Crystal, Reagent, ACS is an inorganic compound that has a pungent salty taste and is soluble water. Ammonium Fluoride is used for p...
Supplier: Honeywell Research Chemicals
Description: Ammonium fluoride, Purity: >/=98%, Grade: Puriss p.a, ACS reagent, Cas number: 12125-01-8, Molecular Formula: NH4F, Molar Mass: 37.0 g/mol, Synonym: Laboratory Chemical, Container: Poly bottle, Size: 100G

SDS

Catalog Number: (BJ52481-50G)
Supplier: Honeywell Research Chemicals
Description: Ammonium fluoride, Purity: >/=98.0% (F), Grade: Analytical, Cas number: 12125-01-8, Molecular Formula: NH4F, Molar mass: 37.0 g/mol, Density: 1.01 g/cm3, Eluent additive for LC-MS, Size: 50G

Catalog Number: (BJ40208H-5L)
Supplier: Honeywell Research Chemicals
Description: We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

SDS


Supplier: BeanTown Chemical
Description: Ammonium fluoride, Grade: ACS, Purity: 98.0% Minimum, CAS Number: 12125-01-8, Molecular Formula: NH4F, Size: 1Kg

SDS

Supplier: BeanTown Chemical
Description: AMMONIUM FLUORIDE 500G

SDS

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM FLUORIDE 1KG

Catalog Number: (BJ40292H-5L)
Supplier: Honeywell Research Chemicals
Description: We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

SDS


Catalog Number: (89400-748)
Supplier: KMG
Description: Ammonium fluoride is used to dilute HF solutions for silicon dioxide film etching.

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM FLUORIDE 500GR

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM FLUORIDE 100GR

Supplier: Strem Chemicals Inc
Description: CAS #: 12125-01-8. Size: 100g.

Supplier: Rigaku Reagents
Description: Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw mate...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom