You Searched For: Ammonium carbonate


17  results were found

SearchResultCount:"17"

Sort Results

List View Easy View

Rate These Search Results

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Ammonium Carbonate, AR® (ACS)
Catalog Number: (97061-374)
Supplier: VWR
Description: AMMONIUM CARBONATE ULTRA PURE GRD 1 KG

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Ammonium Carbonate, BAKER ANALYZED® A.C.S. Reagent
Supplier: MilliporeSigma
Description: AMMONIUM CARBONATE FOR ANALYSI
Supplier: Thermo Scientific Chemicals
Description: Puratronic*, 99.999% (metals basis).Powder,WARNING: Irritates lungs, eyes, skin,50g,250g.
Supplier: Thermo Scientific Chemicals
Description: AMMONIUM CARB NH3 CA 30% 5KG
Catalog Number: (RC622.5-16)
Supplier: Ricca Chemical
Description: AMMONIUM CARBONATE TS 16OZ

Catalog Number: (IC15010880)
Supplier: MP Biomedicals
Description: Ammonium carbonate is used in the washing and defatting of wools, the manufacture of rubber products, in fire extinguishers, and the separation of cac...

SDS


Supplier: Honeywell Research Chemicals
Description: Ammonium carbonate, Purity: >/= 30.0% NH3 basis, Grade: ACS reagent, Cas number: 506-87-6, Molecular Formula: (NH4)2CO3, Molar mass: 96.09 g/mol, Synonym: Hartshorn salt, Laboratory Chemical, ACS Grade Salt, Container: Poly bottle, Appearance: White, Size: 500G
Supplier: Honeywell Research Chemicals
Description: Ammonium carbonate, Puriss, meets analytical specification of NF, Ph. Franc, FCC Hartshorn salt, CAS Number: 506-87-6, Molecular Formula: (NH4)2CO3, Molecular weight: 96.09 g/mol, color: White, Size: 1KG

SDS

Supplier: Spectrum Chemicals
Description: Ammonium Carbonate, Chips, Purified is a white, ammoniacal, solid salt that degrades to gaseous ammonia and carbon dioxide upon heating. It is used as...
Supplier: BeanTown Chemical
Description: Ammonium carbonate, 99.999% trace metals basis, Cas number: 506-87-6, Molecular formula: (NH4)2CO3, Size: 250G

SDS

Supplier: Thermo Scientific Chemicals
Description: In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
Supplier: BeanTown Chemical
Description: AMMONIUM CARBONATE 500G

SDS

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM CARBONATE 1KG

Supplier: Thermo Scientific Chemicals
Description: AMMONIUM CARBONATE1KG

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 17
no targeter for Bottom