You Searched For: Glutaraldehyde


29  results were found

SearchResultCount:"29"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR
Description: GLUTARALDEHYDE 50% SOLN RGT GRD 100ML
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Glutaraldehyde, 25% Aqueous Solution, BAKER ANALYZED® Reagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Description: Glutaraldehyde (25% in H2O), Practical
Catalog Number: (97064-910)
Supplier: VWR
Description: GLUTARALDEHYDE 50% SOL PRTM GRD 500ML

Supplier: MilliporeSigma
Description: GLUTARDIALDEHYDE (25% SOL (1L)
Supplier: MilliporeSigma
Description: Glutardialdehyde is a fixing agent for specimens destined for morphological and enzyme histochemical analysis of samples of human origin.

SDS

Supplier: MilliporeSigma
Description: GLUTARDIALDEHYDE SOLUTION (1L)
Supplier: MilliporeSigma
Description: GLUTARDIALDEHYDE (50% (100ML)
Supplier: MP Biomedicals
Description: Glutaraldehyde is a bifunctional protein cross-linking reagent, reacting with NH2 groups to form Schiff's bases.
Sporicidal agent. Glutaraldehyde...

Supplier: TCI America
Description: CAS Number: 111-30-8 MDL Number: MFCD00007025 Molecular Formula: C5H8O2 Molecular Weight: 100.12 Form: Clear Liquid Color: Colorless Specific Grav...
Supplier: TCI America
Description: CAS Number: 111-30-8 MDL Number: MFCD00007025 Molecular Formula: C5H8O2 Molecular Weight: 100.12 Form: Clear Liquid Color: Colorless Specific Grav...
Catalog Number: (IC19519910)
Supplier: MP Biomedicals
Description: Glutaraldehyde is also referred to as glutaral, 1,5-pentanedione, potentiated acid Glutaraldehyde, sonacide, and glutardialdehyde. Pure monomeric glut...

Supplier: Thermo Scientific Chemicals
Description: 100ML
Catalog Number: (TS11998-0025)
Supplier: Thermo Scientific Chemicals
Description: GLUTARIC DIALDEHYDE 2.5L


Catalog Number: (EM354400-500ML)
Supplier: MilliporeSigma
Description: Form Clear light yellow liquid Cas number 111-30-8 Storage +15C to +30C Used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues.

Supplier: Electron Microscopy Sciences
Description: OCHCH2CH2CH2CHO CAS #111-30-8 Specific Gravity at 20/20[degree]C:1.131 Purified to EM Grade by "all-glass vacuum-distillation." Free from polymers and contaminants. Purity is guaranteed. Sealed under dry nitrogen.

Minority or Woman-Owned Business Enterprise

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 29
no targeter for Bottom