    Sort by:
  • Search Within Results
Alphacel non-nutritive bulk
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibers.
Alphacel Non-N...