Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ZING Green Safety Eco Safety Sign NOTICE, Employees Only, ZING Enterprises

Supplier: ZING Enterprises
1995A 1995S 1995 2995 2995A 2995S
The sign is available in 2 sizes, 7 x 10", and 10 x 14", and three eco-friendly formats.

  • UL Validated for recycled content
  • Made in USA
  • High quality

Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks

Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.

Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.

All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.



 

ORDER

Expand All / Collapse All
Description Material Length Width Supplier No. VWR Catalog Number Unit Price Quantity
ZING Sign NOTICE, Employees Only Plastic 7" 10" 1995 76185-398 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
ZING Sign NOTICE, Employees Only Aluminum 7" 10" 1995A 76185-400 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
ZING Sign NOTICE, Employees Only Self-Adhesive 7" 10" 1995S 76185-402 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
ZING Sign NOTICE, Employees Only Plastic 10" 14" 2995 76185-404 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
ZING Sign NOTICE, Employees Only Aluminum 10" 14" 2995A 76185-406 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
ZING Sign NOTICE, Employees Only Self-Adhesive 10" 14" 2995S 76185-408 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste, Product and/or packaging is safer to human and environmental health
Environmental Certification UL Certified

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping