Y-27632

Supplier: IXCELLS BIOTECHNOLOGIES USA MS

Synonyms: (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide, 2HCl(R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochlorideROCK InhibitorY-27632 dihydrochlorideRho Kinase Inhibitor VI

MD-0025-2MG
103521-494EA 252.57 USD
103521-494
Y-27632
Y-27632

Y-27632 ; 2mg (10 mm in 624.4 µL DMSO); Y-27632 is a selective ROCK1 (p160ROCK) inhibitor with Ki of 140 nm in a cell-free assay, exhibits >200-fold selectivity over other kinases, including PKC, cAMP-dependent protein kinase, MLCK and PAK.

Formula: C₁₄H₂₁N₃O·2HCl
MW: 320.26 g/mol
MDL Number: MFCD03490488
CAS Number: 146986-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR