Avantor® Genesis, C4-300 HPLC Columns
Supplier: VWR
Avantor® Genesis
8P25976
75813-106EA 3365 USD
75813-106
Avantor® Genesis, C4-300 HPLC Columns
Chromatography Columns
Avantor® Genesis phases are based on high purity metal-free spherical silica.
  • Good peak shape and reproducibility
  • Long column lifetime
  • pH stability 1 to 10
They are suitable for the analysis of a wide range of analytes.