Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Promosil Preparative Columns, Bonna-Agela Technologies

Supplier: PHENOMENEX, INC.
PM900220-0 PM901550-0 PM951510-0 PM950220-0 PM951520-0 PM951530-0 PM901510-0 PM901520-0 PM901530-0 PM902510-0 PM902520-0 PM950120-0 PM952510-0 PM951020-0 PM952520-0 PM951030-0 PM952530-0 PM902550-0 PM902530-0 PM950520-0 PM900120-0
Promosil C18 is manufactured with high-purity mono-silane through a proprietary bonding process

  • Good peak shape for both acidic and basic compounds
  • Excellent tolerance to contamination and long lifetime and efficient for high performance-to-cost value

They have high surface bonding coverage and are completely endcapped.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Semi-Preparative C18 10 x 150 10 µm 100 Å PM901510-0 97022-786 Each Retrieving
Semi-Preparative C18 10 x 250 10 µm 100 Å PM902510-0 97022-794 Each Retrieving
Preparative C18 21.2 x 250 10 µm 100 Å PM902520-0 97022-796 Each Retrieving
Preparative C18 30 x 250 10 µm 100 Å PM902530-0 97022-798 Each Retrieving
Preparative C18 21.2 x 150 10 µm 100 Å PM901520-0 97022-788 Each Retrieving
Preparative Guard Cartridge C18 21.2 x 10 5 µm 100 Å PM950120-0 97023-694 Each Retrieving
Preparative Guard Cartridge C18 21.2 x 20 5 µm 100 Å PM950220-0 97023-696 Each Retrieving
Preparative Guard Cartridge C18 21.2 x 20 10 µm 100 Å PM900220-0 97022-784 Each Retrieving
Preparative C18 30 x 150 10 µm 100 Å PM901530-0 97022-790 Each Retrieving
Preparative C18 50 x 250 10 µm 100 Å PM902550-0 97023-692 Each Retrieving
Preparative Guard Cartridge C18 21.2 x 10 10 µm 100 Å PM900120-0 97022-782 Each Retrieving
Preparative C18 21.2 x 100 5 µm 100 Å PM951020-0 97022-818 Each Retrieving
Preparative C18 30 x 150 5 µm 100 Å PM951530-0 97023-706 Each Retrieving
Preparative C18 21.2 x 150 5 µm 100 Å PM951520-0 97022-824 Each Retrieving
Preparative C18 30 x 250 5 µm 100 Å PM952530-0 97023-710 Each Retrieving
Preparative C18 21.2 x 250 5 µm 100 Å PM952520-0 97022-830 Each Retrieving
Preparative C18 50 x 150 10 µm 100 Å PM901550-0 97022-792 Each Retrieving
Preparative C18 21.2 x 50 5 µm 100 Å PM950520-0 97022-812 Each Retrieving
Preparative C18 30 x 100 5 µm 100 Å PM951030-0 97022-820 Each Retrieving
Semi-Preparative C18 10 x 150 5 µm 100 Å PM951510-0 97023-704 Each Retrieving
Semi-Preparative C18 10 x 250 5 µm 100 Å PM952510-0 97022-828 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Carbon Load 18%
Endcapped Double
pH Range 1.5 – 9.0
Surface Area 320 m²/g
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty