Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protein A, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
25282 25284 25283 25286 25285 25288 25287 25289
This protein A provides maximum resolution in two-step labeling.

  • Gold particle sizes of 6, 10 ,15 and 25 nm
  • Monodisperse particle population
  • Minimal size variation and overlap
  • Maximum of one gold particle per primary antibody

The monodisperse size population makes the conjugates suited for multiple labeling with no overlap. The Conventional Immunogold Reagents are the classical conjugates in immuno electron microscopy; they are a good choice when the antigen is abundant and the accessibility of the antigen is relatively good. The conventional labeling approach in transmission and scanning electron microscopy utilizes secondary immunogold reagents based on particles that can be observed without enhancement. These conjugates are suited for single and multiple labeling in electron microscopy, when the number of antigens available for binding is such that a relevant signal can be obtained.

Delivery information: The reagents are supplied in PBS with 1% Bovine Serum Albumin and 15 mM NaN3 at an OD 520 nm of 1.0.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mL 25283 100487-656 Each (1ml) Retrieving Restricted
2.5 mL 25282 100487-654 Each (2,500µl) Retrieving Restricted
1 mL 25285 100487-660 Each (1ml) Retrieving Restricted
2.5 mL 25284 100487-658 Each (1ml) Retrieving Restricted
1.4 mL 25287 100487-664 Each (1ml) Retrieving Restricted
3.5 mL 25286 100487-662 Each (1ml) Retrieving Restricted
1.4 mL 25289 100487-668 Each (1ml) Retrieving Restricted
3.5 mL 25288 100487-666 Each (1ml) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name EM Protein-A/Gold, 10 nm EM Protein-A/Gold, 15 nm EM Protein-A/Gold, 25 nm EM Protein-A/Gold, 15 nm EM Protein-A/Gold, 25 nm EM Protein-A/Gold, 6 nm EM Protein-A/Gold, 10 nm EM Protein-A/Gold, 6 nm
Grade EM

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping